ENA018106517 Enamine

2-(2-methoxyethoxy)-5-(trifluoromethyl)aniline

  • MDL number MFCD03966847
  • Cas Number : 49750-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-42
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02941
Availability details for
Quantity
  • Availability
  • Add to Cart