ENA018106522 Enamine

2-(4-ethoxyphenoxy)-5-(trifluoromethyl)aniline

  • MDL number MFCD03966896
  • Cas Number : 565166-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-46
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02946
Availability details for
Quantity
  • Availability
  • Add to Cart