ENA018106540 Enamine

6-Amino-1-benzyl-3-ethyl-1H-pyrimidine-2,4-dione

  • MDL number MFCD03658006
  • Cas Number : 749219-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02968

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02968
Availability details for
Quantity
  • Availability
  • Add to Cart