ENA018106544 Enamine

5-(adamantan-1-yl)-1,3,4-thiadiazol-2-amine

  • Cas Number : 26526-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02972
Availability details for
Quantity
  • Availability
  • Add to Cart