ENA018106546 Enamine

3-(4-Naphthalen-2-yl-thiazol-2-yl)-pyridine

  • MDL number MFCD03479852
  • Cas Number : 519151-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02974

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02974
Availability details for
Quantity
  • Availability
  • Add to Cart