ENA018106547 Enamine

3-[4-(2,3,4-Trichloro-phenyl)-thiazol-2-yl]-pyridine

  • MDL number MFCD03481657
  • Cas Number : 521280-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02975

Safety Information

msds https://www.enaminestore.com/catalog/EN300-02975
Availability details for
Quantity
  • Availability
  • Add to Cart