ENA018106561 Enamine

5-phenyl-3-(prop-2-en-1-yl)-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD03815832
  • Cas Number : 301233-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-02998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02998
Availability details for
Quantity
  • Availability
  • Add to Cart