ENA018106569 Enamine

2-{1-[4-(dimethylamino)phenyl]-2,5-dimethyl-1H-pyrrol-3-yl}-1,1,1,3,3,3-hexafluoropropan-2-ol

  • MDL number MFCD02696590
  • Cas Number : 328110-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03006
Availability details for
Quantity
  • Availability
  • Add to Cart