ENA018106572 Enamine

N-(2-Hydroxy-ethyl)-3-(morpholine-4-sulfonyl)-benzamide

  • MDL number MFCD03147246
  • Cas Number : 379254-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03009

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03009
Availability details for
Quantity
  • Availability
  • Add to Cart