ENA018106578 Enamine

{6-chloroimidazo[2,1-b][1,3]thiazol-5-yl}methanol

  • MDL number MFCD02253174
  • Cas Number : 24918-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-148
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03016
Availability details for
Quantity
  • Availability
  • Add to Cart