ENA018106587 Enamine

5-(2,4-dichlorophenyl)-4-[2-(propan-2-yl)phenyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02725624
  • Cas Number : 380345-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03025
Availability details for
Quantity
  • Availability
  • Add to Cart