ENA018106591 Enamine

5-[(2,3-dimethylphenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD03147286
  • Cas Number : 379254-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-196
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03029
Availability details for
Quantity
  • Availability
  • Add to Cart