ENA018106593 Enamine

3-(2,4-dimethylphenyl)-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD02720534
  • Cas Number : 22458-49-7
  • Molecular Weight : 282.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03031
Availability details for
Quantity
  • Availability
  • Add to Cart