ENA018106596 Enamine

5-[(2-methylphenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD00125833
  • Cas Number : 27385-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03034
Availability details for
Quantity
  • Availability
  • Add to Cart