ENA018106597 Enamine

5-[(3,4-dimethylphenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD02720739
  • Cas Number : 380191-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-182
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03035
Availability details for
Quantity
  • Availability
  • Add to Cart