ENA018106604 Enamine

3-[4-(propan-2-yl)phenyl]-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD02724567
  • Cas Number : 380342-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03042
Availability details for
Quantity
  • Availability
  • Add to Cart