ENA018106617 Enamine

4-(2,4-difluorophenyl)-5-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02720698
  • Cas Number : 380189-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03055
Availability details for
Quantity
  • Availability
  • Add to Cart