ENA018106618 Enamine

5-[(4-methylphenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD00125725
  • Cas Number : 14731-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-192
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03056
Availability details for
Quantity
  • Availability
  • Add to Cart