ENA018106621 Enamine

4-(3-Chloro-2-methyl-phenyl)-5-pyridin-4-yl-4H-[1,2,4]triazole-3-thiol

  • MDL number MFCD02724835
  • Cas Number : 380335-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 322...324
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03059

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03059
Availability details for
Quantity
  • Availability
  • Add to Cart