ENA018106623 Enamine

5-{[4-(propan-2-yl)phenyl]amino}-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD02724842
  • Cas Number : 380335-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03061
Availability details for
Quantity
  • Availability
  • Add to Cart