ENA018106632 Enamine

2-(1,3-benzothiazol-2-yl)-3-[(3,4-dimethylphenyl)amino]-3-sulfanylprop-2-enenitrile

  • Cas Number : 379255-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03070
Availability details for
Quantity
  • Availability
  • Add to Cart