ENA018106646 Enamine

3-(2-Chloro-6-fluoro-phenyl)-7-hydroxy-2-methyl-chromen-4-one

  • MDL number MFCD02714411
  • Cas Number : 377769-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220...222
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03084

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03084
Availability details for
Quantity
  • Availability
  • Add to Cart