ENA018106649 Enamine

3-(Biphenyl-4-yloxy)-7-hydroxy-2-methyl-chromen-4-one

  • MDL number MFCD02725378
  • Cas Number : 380348-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03087

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03087
Availability details for
Quantity
  • Availability
  • Add to Cart