ENA018106653 Enamine

ethyl 5-hydroxy-2-phenyl-1-benzofuran-3-carboxylate

  • MDL number MFCD00631149
  • Cas Number : 4610-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-156
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03091
Availability details for
Quantity
  • Availability
  • Add to Cart