ENA018106658 Enamine

2-Mercapto-benzooxazole-5-sulfonic acid (4-methoxy-phenyl)-amide

  • MDL number MFCD03042005
  • Cas Number : 380347-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03097

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03097
Availability details for
Quantity
  • Availability
  • Add to Cart