ENA018106676 Enamine

ethyl 1-ethyl-2-sulfanyl-1H-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD00435402
  • Cas Number : 295345-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-153
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03116
Availability details for
Quantity
  • Availability
  • Add to Cart