ENA018106680 Enamine

5-(4-chlorophenyl)-4-(2-methylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD01654773
  • Cas Number : 309274-06-4
  • Molecular Weight : 301.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03120
Availability details for
Quantity
  • Availability
  • Add to Cart