ENA018106682 Enamine

5-[(4-phenoxyphenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD03150680
  • Cas Number : 380431-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-184
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03122
Availability details for
Quantity
  • Availability
  • Add to Cart