ENA018106683 Enamine

4-phenyl-5-[3-(pyrrolidine-1-sulfonyl)phenyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03150684
  • Cas Number : 380431-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03123
Availability details for
Quantity
  • Availability
  • Add to Cart