ENA018106695 Enamine

4-(2-methoxy-5-methylphenyl)-5-(thiophen-2-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03152702
  • Cas Number : 379728-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03135
Availability details for
Quantity
  • Availability
  • Add to Cart