ENA018106696 Enamine

1-(4-butylphenyl)-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD03154352
  • Cas Number : 135965-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-84
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03136
Availability details for
Quantity
  • Availability
  • Add to Cart