ENA018106702 Enamine

3-(2-ethoxyphenyl)-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD03152700
  • Cas Number : 65141-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03142
Availability details for
Quantity
  • Availability
  • Add to Cart