ENA018106732 Enamine

5-(2,3-dihydro-1,4-benzodioxin-2-yl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD00455502
  • Cas Number : 120423-45-2
  • Molecular Weight : 236.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03177
Availability details for
Quantity
  • Availability
  • Add to Cart