ENA018106733 Enamine

5-(3-chloro-1-benzothiophen-2-yl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD03964652
  • Cas Number : 144605-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03178
Availability details for
Quantity
  • Availability
  • Add to Cart