ENA018106743 Enamine

5-(1H-indol-3-yl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD03965935
  • Cas Number : 379725-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-225
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03212
Availability details for
Quantity
  • Availability
  • Add to Cart