ENA018106749 Enamine

3-(4-chlorophenoxy)-7-hydroxy-4H-chromen-4-one

  • MDL number MFCD00525545
  • Cas Number : 137987-94-1
  • Molecular Weight : 288.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 265-266
Iupac Name 3-(4-chlorophenoxy)-7-hydroxy-4H-chromen-4-one
Inchi Code 1S/C15H9ClO4/c16-9-1-4-11(5-2-9)20-14-8-19-13-7-10(17)3-6-12(13)15(14)18/h1-8,17H
Inchi Key BIWKGGSODHYGIP-UHFFFAOYSA-N

Description

Melting Point 265-266
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03224
Availability details for
Quantity
  • Availability
  • Add to Cart