ENA018106766 Enamine

7-hydroxy-3-(4-methoxyphenyl)-2-methyl-4H-chromen-4-one

  • MDL number MFCD01995502
  • Cas Number : 13004-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-03241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03241
Availability details for
Quantity
  • Availability
  • Add to Cart