ENA018106784 Enamine

7-hydroxy-3-(1-phenyl-1H-pyrazol-4-yl)-4H-chromen-4-one

  • MDL number MFCD01235440
  • Cas Number : 61034-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03261
Availability details for
Quantity
  • Availability
  • Add to Cart