ENA018106794 Enamine

5-(4-methoxyphenyl)-4-(2-methylpropyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03478379
  • Cas Number : 438031-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03271
Availability details for
Quantity
  • Availability
  • Add to Cart