ENA018106795 Enamine

4-(2-methoxyethyl)-5-(pyridin-4-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03414048
  • Cas Number : 663181-77-9
  • Molecular Weight : 236.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03272
Availability details for
Quantity
  • Availability
  • Add to Cart