ENA018106799 Enamine

5-(4-chlorophenyl)-4-(2-methoxyethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03966921
  • Cas Number : 26438-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03276
Availability details for
Quantity
  • Availability
  • Add to Cart