ENA018106800 Enamine

4-(2-methylphenyl)-5-(pyridin-4-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03651673
  • Cas Number : 16629-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03277
Availability details for
Quantity
  • Availability
  • Add to Cart