ENA018106803 Enamine

5-[(4-ethoxyphenyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD02956713
  • Cas Number : 68161-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03280
Availability details for
Quantity
  • Availability
  • Add to Cart