ENA018106808 Enamine

4-(4-ethoxyphenyl)-5-(1H-indol-3-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03955631
  • Cas Number : 885458-58-2
  • Molecular Weight : 336.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03285
Availability details for
Quantity
  • Availability
  • Add to Cart