ENA018106816 Enamine

2-Mercapto-3-(2-methoxy-ethyl)-3H-quinazolin-4-one

  • MDL number MFCD01567624
  • Cas Number : 422273-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03293

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03293
Availability details for
Quantity
  • Availability
  • Add to Cart