ENA018106829 Enamine

4-(3,4-dichlorophenyl)-5-(pyridin-4-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03478375
  • Cas Number : 438031-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03306
Availability details for
Quantity
  • Availability
  • Add to Cart