ENA018106833 Enamine

4-(3-methylphenyl)-5-sulfanyl-8-thia-4,6-diazatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),5-trien-3-one

  • MDL number MFCD03284223
  • Cas Number : 341499-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03310
Availability details for
Quantity
  • Availability
  • Add to Cart