ENA018106834 Enamine

11-(2-methylphenyl)-10-sulfanyl-7-thia-9,11-diazatricyclo[6.4.0.0,?]dodeca-1(8),2(6),9-trien-12-one

  • MDL number MFCD02990657
  • Cas Number : 380583-57-3
  • Molecular Weight : 314.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03311
Availability details for
Quantity
  • Availability
  • Add to Cart