ENA018106841 Enamine

4-amino-5-(pyridin-3-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00834356
  • Cas Number : 78027-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-211
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03318
Availability details for
Quantity
  • Availability
  • Add to Cart