ENA018106842 Enamine

3-(9-ethyl-9H-carbazol-3-yl)-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD03478329
  • Cas Number : 438030-15-0
  • Molecular Weight : 371.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-03319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03319
Availability details for
Quantity
  • Availability
  • Add to Cart